Perfluorophenanthrene ≥80%

Supplier: Matrix Scientific

Synonyms: Perfluoroperhydrophenanthrene

001099-100G
101780-952EA 369.23 USD
101780-952
Perfluorophenanthrene ≥80%
Perfluorophenanthrene

TCA listed: T
Hazard: Irritant

Formula: C₁₄F₂₄
MW: 624.116 g/mol
Boiling Pt: 215 °C
Melting Pt: –20 °C
Density: 2.03
MDL Number: MFCD00042596
CAS Number: 306-91-2

Order Now

Specification Test Results

Boiling point 215°C
Melting point -20°C
Density 2.03
ND20 1.3348

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR