3-(Trifluoromethyl)quinoxaline-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
070809-500MG
070809-1G
101946-414EA
803.08
USD
101946-414
101946-416
3-(Trifluoromethyl)quinoxaline-2-carboxylic acid ≥95%
3-(Trifluoromethyl)quinoxaline-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₅F₃N₂O₂ Melting Pt: 164 °C (dec.) |
MDL Number:
MFCD11869266 CAS Number: 1142190-60-0 |
Specification Test Results
Melting point | 164°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...