tert-Butyl-4-(5-bromo-1,3,4-thiadiazol-2-yl)piperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

070804-500MG
101946-396EA 515.38 USD
101946-396
tert-Butyl-4-(5-bromo-1,3,4-thiadiazol-2-yl)piperazine-1-carboxylate ≥97%
tert-Butyl-4-(5-bromo-1,3,4-thiadiazol-2-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇BrN₄O₂S
MW: 349.26 g/mol
Melting Pt: 95…97 °C
MDL Number: MFCD12923104
CAS Number: 1352925-43-9

Order Now

Specification Test Results

Melting point 95-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR