7H-Pyrrolo[2,3-d]pyrimidine-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
070803-500MG
101946-394EA
538.46
USD
101946-394
7H-Pyrrolo[2,3-d]pyrimidine-4-carboxylic acid ≥95%
7H-Pyrrolo[2,3-d]pyrimidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₇H₅N₃O₂ Melting Pt: 215 °C (dec.) |
MDL Number:
MFCD13193623 CAS Number: 1005206-17-6 |
Specification Test Results
Melting point | 215°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...