Methyl-2-amino-4-fluoro-5-methoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-4-fluoro-5-methoxybenzoate

070794-500MG 070794-1G
101946-354EA 803.08 USD
101946-354 101946-356
Methyl-2-amino-4-fluoro-5-methoxybenzoate ≥95%
Methyl-2-amino-4-fluoro-5-methoxybenzoate

Hazard: Irritant

Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Melting Pt: 43…46 °C
MDL Number: MFCD16159159
CAS Number: 159768-51-1

Order Now

Specification Test Results

Melting point 43-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR