6-Methoxy-5-methyl-2,3-dihydro-1H-isoindol-1-one ≥95%

Supplier: Matrix Scientific

070791-1G 070791-500MG
101946-344EA 692.31 USD
101946-344 101946-342
6-Methoxy-5-methyl-2,3-dihydro-1H-isoindol-1-one ≥95%
6-Methoxy-5-methyl-2,3-dihydro-1H-isoindol-1-one

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
Melting Pt: 164…166 °C
MDL Number: MFCD12924304
CAS Number: 1138220-76-4

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR