4-(4-Ethyl-1H-1,2,3-triazol-1-yl)aniline ≥95%

Supplier: Matrix Scientific

070787-500MG
101946-328EA 803.08 USD
101946-328
4-(4-Ethyl-1H-1,2,3-triazol-1-yl)aniline ≥95%
4-(4-Ethyl-1H-1,2,3-triazol-1-yl)aniline

Hazard: Irritant

Formula: C₁₀H₁₂N₄
Melting Pt: 104…106 °C
MDL Number: MFCD21333209
CAS Number: 1357147-47-7

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR