Ethyl-2,6-bis(trifluoromethyl)pyridine-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2,6-bis(trifluoromethyl)isonicotinate
070783-500MG
070783-1G
101946-314EA
1743.08
USD
101946-314
101946-312
Ethyl-2,6-bis(trifluoromethyl)pyridine-4-carboxylate ≥95%
Ethyl-2,6-bis(trifluoromethyl)pyridine-4-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇F₆NO₂ MW: 287.16 g/mol Melting Pt: 41…43 °C |
MDL Number:
MFCD17015230 CAS Number: 1280520-88-8 |
Specification Test Results
Melting point | 41-43°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...