4,4-Dimethyl-1,2-oxazolidin-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4,4-Dimethylisoxazolidin-3-one

070782-500MG 070782-1G
101946-310EA 846.15 USD
101946-310 101946-308
4,4-Dimethyl-1,2-oxazolidin-3-one ≥95%
4,4-Dimethyl-1,2-oxazolidin-3-one

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 74…76 °C
MDL Number: MFCD21333197
CAS Number: 81778-07-6

Order Now

Specification Test Results

Melting point 74-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR