2-Chloro-3-(trifluoromethyl)quinoxaline ≥95%
Supplier: Matrix Scientific
070779-1G
101946-296EA
292.31
USD
101946-296
2-Chloro-3-(trifluoromethyl)quinoxaline ≥95%
2-Chloro-3-(trifluoromethyl)quinoxaline
Hazard: Irritant
Formula:
C₉H₄ClF₃N₂ MW: 232.59 g/mol Melting Pt: 147…149 °C |
MDL Number:
MFCD00243891 CAS Number: 254732-51-9 |
Specification Test Results
Melting point | 147-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...