1-(5-Bromo-1,3,4-thiadiazol-2-yl)piperazine ≥97%

Supplier: Matrix Scientific

070775-500MG
101946-282EA 995.38 USD
101946-282
1-(5-Bromo-1,3,4-thiadiazol-2-yl)piperazine ≥97%
1-(5-Bromo-1,3,4-thiadiazol-2-yl)piperazine

Hazard: Irritant

Formula: C₆H₉BrN₄S
Melting Pt: 111…113 °C
MDL Number: MFCD20369138
CAS Number: 1340396-26-0

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR