4-(5-Bromo-1,3,4-thiadiazol-2-yl)-cis-2,6-dimethylmorpholine ≥95%

Supplier: Matrix Scientific

070771-500MG
101946-274EA 803.08 USD
101946-274
4-(5-Bromo-1,3,4-thiadiazol-2-yl)-cis-2,6-dimethylmorpholine ≥95%
4-(5-Bromo-1,3,4-thiadiazol-2-yl)-cis-2,6-dimethylmorpholine

Hazard: Irritant

Formula: C₈H₁₂BrN₃OS
Melting Pt: 69…71 °C
MDL Number: MFCD21333210
CAS Number: 1357147-40-0

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR