2-Bromo-5-(pyrrolidin-1-yl)-1,3,4-thiadiazole ≥97%

Supplier: Matrix Scientific

070770-500MG
101946-272EA 803.08 USD
101946-272
2-Bromo-5-(pyrrolidin-1-yl)-1,3,4-thiadiazole ≥97%
2-Bromo-5-(pyrrolidin-1-yl)-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₆H₈BrN₃S
Melting Pt: 110…112 °C
MDL Number: MFCD20368503
CAS Number: 1339586-51-4

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR