tert-Butyl-4-[6-(ethoxycarbonyl)pyridin-2-yl]piperazine-1-carboxylate

Supplier: Matrix Scientific

067210-500MG 067210-1G
101933-540EA 803.08 USD
101933-540 101933-542
tert-Butyl-4-[6-(ethoxycarbonyl)pyridin-2-yl]piperazine-1-carboxylate
tert-Butyl-4-[6-(ethoxycarbonyl)pyridin-2-yl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₄
MW: 335.41 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD20921603
CAS Number: 1350807-20-3

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR