(-)-Di-p-toluoyl-L-tartaric acid ≥98%
Supplier: Matrix Scientific
Synonyms:
(2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid
, Di-p-toluoyl-L-tartaric acid
, (-)-O,O'-Di-p-toluoyl-L-tartaric acid
067066-100G
067066-25G
101933-118EA
116.92
USD
101933-118
101933-116
(-)-Di-p-toluoyl-L-tartaric acid ≥98%
(-)-Di-p-toluoyl-L-tartaric acid
TCA listed: T
Hazard: Irritant
Formula:
C₂₀H₁₈O₈ MW: 386.36 g/mol Melting Pt: 169…171 °C |
MDL Number:
MFCD00064440 CAS Number: 32634-66-5 |
Specification Test Results
Melting point | 169-171°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...