Methyl-4-(morpholin-4-ylmethyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(morpholinomethyl)benzoate

066928-500MG 066928-1G
101932-784EA 403.08 USD
101932-784 101932-786
Methyl-4-(morpholin-4-ylmethyl)benzoate ≥95%
Methyl-4-(morpholin-4-ylmethyl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MW: 235.29 g/mol
MDL Number: MFCD01167691
CAS Number: 68453-56-5

Order Now

Specification Test Results

Melting point Barelygree

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR