2-Fluoro-4-nitrobenzaldehyde ≥97%

Supplier: Matrix Scientific

066799-1G
101932-398EA 59.8 USD
101932-398
2-Fluoro-4-nitrobenzaldehyde ≥97%
2-Fluoro-4-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄FNO₃
MW: 169.10 g/mol
Melting Pt: 98…99 °C
MDL Number: MFCD00968941
CAS Number: 157701-72-9

Order Now

Specification Test Results

Melting point 98-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR