Triphenylmethyl isothiocyanate

Supplier: Matrix Scientific

057211-25G
101912-768EA 915.38 USD
101912-768
Triphenylmethyl isothiocyanate
Triphenylmethyl isothiocyanate

Hazard: Toxic

Formula: C₂₀H₁₅NS
Melting Pt: 138…140 °C
MDL Number: MFCD00014443
CAS Number: 1726-94-9

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR