Terephthalic acid dihydrazide

Supplier: Matrix Scientific

057188-10G
101912-690EA 85 USD
101912-690
Terephthalic acid dihydrazide
Terephthalic acid dihydrazide

CAS: 136-64-1
MDL Number: MFCD00014762
Molecular Formula: C8H10N4O2
Molecular Weight: 194,19
Hazard: Irritant

Formula: C₈H₁₀N₄O₂
MW: 194,19
Melting Pt: >300 °C (dec.)
MDL Number: MFCD00014762
CAS Number: 136-64-1
EINECS: 205-253-2

Order Now

Specification Test Results

Melting point 300°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR