Stearic acid hydrazide

Supplier: Matrix Scientific

057187-50G
101912-686EA 155.25 USD
101912-686
Stearic acid hydrazide
Stearic acid hydrazide

Hazard: Irritant

Formula: C₁₈H₃₈N₂O
Melting Pt: 112…115 °C
MDL Number: MFCD00014764
CAS Number: 4130-54-5

Order Now

Specification Test Results

Melting point 112-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR