4-Phenylbenzhydrazide

Supplier: Matrix Scientific

Synonyms: 4-Biphenylcarboxylic acid hydrazide

057176-50G
101912-646EA 807.69 USD
101912-646
4-Phenylbenzhydrazide
4-Phenylbenzhydrazide

Hazard: Irritant

Formula: C₁₃H₁₂N₂O
MW: 212.2472 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00017078
CAS Number: 18622-23-6

Order Now

Specification Test Results

Melting point 189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR