3-(4-Phenoxyphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

057175-5G
101912-642EA 449.23 USD
101912-642
3-(4-Phenoxyphenyl)propionic acid ≥98%
3-(4-Phenoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
Melting Pt: 84…86 °C
MDL Number: MFCD00220674
CAS Number: 20062-91-3

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR