3-(3-Phenoxyphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

057174-5G
101912-638EA 461.54 USD
101912-638
3-(3-Phenoxyphenyl)propionic acid ≥98%
3-(3-Phenoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
Melting Pt: 62…64 °C
MDL Number: MFCD06823981
CAS Number: 52888-70-7

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR