Pentamethylphenylacetonitrile ≥99%

Supplier: Matrix Scientific

057168-5G
101912-614EA 272.31 USD
101912-614
Pentamethylphenylacetonitrile ≥99%
Pentamethylphenylacetonitrile

Hazard: Irritant

Formula: C₁₃H₁₇N
Melting Pt: 113…115 °C
MDL Number: MFCD00045622
CAS Number: 34688-70-5

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR