4-Morpholinethiocarboxamide

Synonyms: Morpholine-4-carbothioamide

057155-10G
101912-562EA 620.34 USD
101912-562
4-Morpholinethiocarboxamide
4-Morpholinethiocarboxamide

Hazard: Irritant

Formula: C₅H₁₀N₂OS
MW: 146.21 g/mol
Melting Pt: 169…170 °C
MDL Number: MFCD00178434
CAS Number: 14294-10-1

Order Now

Specification Test Results

Melting point 169-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR