3-Methyl-2-nitrobenzhydrazide

Supplier: Matrix Scientific

057142-50G
101912-522EA 781.54 USD
101912-522
3-Methyl-2-nitrobenzhydrazide
3-Methyl-2-nitrobenzhydrazide

Hazard: Irritant

Formula: C₈H₉N₃O₃
Melting Pt: 152…154 °C
MDL Number: MFCD03425694
CAS Number: 1016745-70-2

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR