3-(3,4-Methylenedioxyphenyl)propionic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
3-(1,3-Benzodioxyl-5-yl)propionic acid
, 3,4-(Methylenedioxy)hydrocinnamic acid
057139-25G
101912-510EA
623.08
USD
101912-510
3-(3,4-Methylenedioxyphenyl)propionic acid ≥98%
3-(3,4-Methylenedioxyphenyl)propionic acid
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Melting Pt: 87…88 °C |
MDL Number:
MFCD00016909 CAS Number: 2815-95-4 |
Specification Test Results
Melting point | 87-88°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...