2-Methoxybiphenyl-3-isothiocyanate

Supplier: Matrix Scientific

Synonyms: (2-Methoxy-5-phenyl)phenyl isothiocyanate

057126-25G
101912-458EA 629.23 USD
101912-458
2-Methoxybiphenyl-3-isothiocyanate
2-Methoxybiphenyl 3-isothiocyanate

Hazard: Irritant

Formula: C₁₄H₁₁NOS
MW: 241.31
Melting Pt: 80…82 °C
MDL Number: MFCD00041078
CAS Number: 206761-68-4

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR