5-(4-Iodophenyl)-1H-tetrazole ≥95%

Supplier: Matrix Scientific

057106-1G
101912-396EA 769.23 USD
101912-396
5-(4-Iodophenyl)-1H-tetrazole ≥95%
5-(4-Iodophenyl)-1H-tetrazole

Hazard: Irritant

Formula: C₇H₅IN₄
Melting Pt: 269…271 °C (dec.)
MDL Number: MFCD09025698
CAS Number: 53324-36-0

Order Now

Specification Test Results

Melting point 269-271°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR