3-Hydroxy-o-toluic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-methylbenzoic acid

057095-25G
101912-352EA 41.54 USD
101912-352
3-Hydroxy-o-toluic acid ≥97%
3-Hydroxy-o-toluic acid

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD00671541
CAS Number: 603-80-5

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR