2-Hydroxy-3-methylbenzhydrazide

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-3-methylbenzohydrazide

057094-5G
101912-346EA 166.75 USD
101912-346
2-Hydroxy-3-methylbenzhydrazide
2-Hydroxy-3-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 190…191 °C
MDL Number: MFCD00017052
CAS Number: 30991-42-5

Order Now

Specification Test Results

Melting point 190-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR