(-)-4'-Fluorotartranilic acid

Supplier: Matrix Scientific

057087-5G
101912-322EA 412.31 USD
101912-322
(-)-4'-Fluorotartranilic acid
(-)-4'-Fluorotartranilic acid

Hazard: Irritant

Formula: C₁₀H₁₀FNO₅
Melting Pt: 192…193 °C (dec.)
MDL Number: MFCD00070633
CAS Number: 238401-54-2

Order Now

Specification Test Results

Melting point 192-193°C (

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR