3-(4-Ethoxy-3-methoxyphenyl)propionic acid ≥98%
Supplier: Matrix Scientific
057066-25G
057066-5G
101912-236EA
896.92
USD
101912-236
101912-234
3-(4-Ethoxy-3-methoxyphenyl)propionic acid ≥98%
3-(4-Ethoxy-3-methoxyphenyl)propionic acid
Hazard: Irritant
Formula:
C₁₂H₁₆O₄ Melting Pt: 130…132 °C |
MDL Number:
MFCD06823980 CAS Number: 30044-91-8 |
Specification Test Results
Melting point | 130-132°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...