4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
Supplier: Matrix Scientific
057056-2G
057056-10G
101912-204EA
470.77
USD
101912-204
101912-202
4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
Hazard: Irritant
Formula:
C₉H₇F₆N₃S MW: 303.23 g/mol Melting Pt: 158…160 °C |
MDL Number:
MFCD00041292 CAS Number: 38901-31-4 |
Specification Test Results
Melting point | 158-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...