3-(2,5-Dimethylphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

057044-25G
101912-160EA 930.77 USD
101912-160
3-(2,5-Dimethylphenyl)propionic acid ≥98%
3-(2,5-Dimethylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₂
Melting Pt: 44…46 °C
MDL Number: MFCD06823972
CAS Number: 25173-75-5

Order Now

Specification Test Results

Melting point 44-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR