2,5-Dimethyl-4-methoxybenzoic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Methoxy-2,5-dimethylbenzoic acid
057037-10G
057037-2G
101912-132EA
458.46
USD
101912-132
101912-130
2,5-Dimethyl-4-methoxybenzoic acid ≥98%
2,5-Dimethyl-4-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₂O₃ MW: 180.21 g/mol Melting Pt: 166…168 °C |
MDL Number:
MFCD01194285 CAS Number: 58106-26-6 |
Specification Test Results
Melting point | 166-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...