3-(2,4-Difluorophenyl)propionic acid
Supplier: Matrix Scientific
057018-5G
057018-25G
101912-062EA
932.31
USD
101912-062
101912-060
3-(2,4-Difluorophenyl)propionic acid
3-(2,4-Difluorophenyl)propionic acid
Hazard: Irritant
Formula:
C₉H₈F₂O₂ Melting Pt: 109…111 °C |
MDL Number:
MFCD04116052 CAS Number: 134672-70-1 |
Specification Test Results
Melting point | 109-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...