1-(5-Chloro-2-methoxyphenyl)-2-thiourea
Supplier: Matrix Scientific
Synonyms:
1-(5-Chloro-2-methoxyphenyl)thiourea
056959-50G
056959-10G
101911-878EA
198.46
USD
101911-878
101911-880
1-(5-Chloro-2-methoxyphenyl)-2-thiourea
1-(5-Chloro-2-methoxyphenyl)-2-thiourea
Hazard: Irritant
Formula:
C₈H₉ClN₂OS MW: 216.69 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD00041169 CAS Number: 63980-69-8 |
Specification Test Results
Melting point | 145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...