2-Chloro-5-iodobenzhydrazide

Supplier: Matrix Scientific

056953-2G 056953-10G
101911-854EA 566.15 USD
101911-854 101911-852
2-Chloro-5-iodobenzhydrazide
2-Chloro-5-iodobenzhydrazide

Hazard: Irritant

Formula: C₇H₆ClIN₂O
Melting Pt: 176…178 °C
MDL Number: MFCD03425676
CAS Number: 1021990-78-2

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR