1-(4-Chlorobenzyl)-2-thiourea

Supplier: Matrix Scientific

056951-10G 056951-2G
101911-846EA 73.85 USD
101911-846 101911-848
1-(4-Chlorobenzyl)-2-thiourea
1-(4-Chlorobenzyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₉ClN₂S
Melting Pt: 142…144 °C
MDL Number: MFCD00060466
CAS Number: 24827-37-0

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR