1-(2-Bromo-4-methylphenyl)-2-thiourea

Supplier: Matrix Scientific

056925-10G 056925-2G
101911-754EA 375.38 USD
101911-754 101911-752
1-(2-Bromo-4-methylphenyl)-2-thiourea
1-(2-Bromo-4-methylphenyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₉BrN₂S
Melting Pt: 174…176 °C
MDL Number: MFCD00041147
CAS Number: 66644-79-9

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR