2-Bromo-3-methylbenzhydrazide

Supplier: Matrix Scientific

056917-1G 056917-5G
101911-722EA 181.54 USD
101911-722 101911-724
2-Bromo-3-methylbenzhydrazide
2-Bromo-3-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₉BrN₂O
Melting Pt: 144…146 °C
MDL Number: MFCD03425673
CAS Number: 1023584-30-6

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR