1-(4-Acetylphenyl)-2-thiourea ≥98%

Supplier: Matrix Scientific

Synonyms: N-(4-Acetylphenyl)thiourea

056877-5G 056877-25G
101911-568EA 307.69 USD
101911-568 101911-570
1-(4-Acetylphenyl)-2-thiourea ≥98%
1-(4-Acetylphenyl)-2-thiourea

Hazard: Irritant

Order Now

Specification Test Results

Melting point 215°C (dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR