4-(3,4,5-Trimethoxyphenyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056833-1G
101911-494EA 106.15 USD
101911-494
4-(3,4,5-Trimethoxyphenyl)-3-thiosemicarbazide
4-(3,4,5-Trimethoxyphenyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₃S
Melting Pt: 178…179 °C
MDL Number: MFCD00060574
CAS Number: 206762-46-1

Order Now

Specification Test Results

Melting point 178-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR