4-(2-Tetrahydrofurfuryl)-3-thiosemicarbazide ≥98%

Supplier: Matrix Scientific

056826-1G
101911-480EA 101.54 USD
101911-480
4-(2-Tetrahydrofurfuryl)-3-thiosemicarbazide ≥98%
4-(2-Tetrahydrofurfuryl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₆H₁₃N₃OS
Melting Pt: 128…130 °C
MDL Number: MFCD00060497
CAS Number: 151672-39-8

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR