Sebacic acid dihydrazide
Supplier: Matrix Scientific
Synonyms:
Decanedihydrazide
056825-25G
101911-478EA
146.15
USD
101911-478
Sebacic acid dihydrazide
Sebacic acid dihydrazide
Hazard: Irritant
Formula:
C₁₀H₂₂N₄O₂ MW: 230.3073 g/mol Melting Pt: 184…185 °C |
MDL Number:
MFCD00039763 CAS Number: 925-83-7 |
Specification Test Results
Melting point | 184-185°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...