1-(2-Piperidinoethyl)-2-thiourea

Supplier: Matrix Scientific

056819-1G
101911-466EA 124.62 USD
101911-466
1-(2-Piperidinoethyl)-2-thiourea
1-(2-Piperidinoethyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₁₇N₃S
Melting Pt: 111…113 °C
MDL Number: MFCD00070655
CAS Number: 206761-87-7

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR