α-Phthalimidopropiophenone ≥97%

Supplier: Matrix Scientific

056815-10G
101911-456EA 461.54 USD
101911-456
α-Phthalimidopropiophenone ≥97%
α-Phthalimidopropiophenone

Hazard: Irritant

Formula: C₁₇H₁₃NO₃
Melting Pt: 87…88 °C
MDL Number: MFCD00060520
CAS Number: 19437-20-8

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR