1-(3-Phenylpropyl)-2-thiourea

Supplier: Matrix Scientific

056811-1G
101911-448EA 93.85 USD
101911-448
1-(3-Phenylpropyl)-2-thiourea
1-(3-Phenylpropyl)-2-thiourea

Hazard: Irritant

Formula: C₁₀H₁₄N₂S
Melting Pt: 111 °C
MDL Number: MFCD00060471
CAS Number: 93168-20-8

Order Now

Specification Test Results

Melting point 111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR