1,3-Phenylenebis(2-thiourea)

Supplier: Matrix Scientific

056808-5G
101911-442EA 116.92 USD
101911-442
1,3-Phenylenebis(2-thiourea)
1,3-Phenylenebis(2-thiourea)

Hazard: Irritant

Formula: C₈H₁₀N₄S₂
Melting Pt: 211 °C
MDL Number: MFCD00060445
CAS Number: 2591-01-7

Order Now

Specification Test Results

Melting point 211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR